Anti-Copper-metallothionein Rabbit Recombinant Polyclonal Antibody (Biotinylated)

Supplier: AFG Bioscience
A51095-100UL A51095-50UL
77816-527EA 521.4 CAD
77816-527 77816-528
Anti-Copper-metallothionein Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Antibodies
Type:
Antigen: Copper-metallothionein
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Roman snail
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR