CELLSTAR® OneWell™ and FourWell™ Plates, Greiner Bio-One
Supplier: Greiner Bio-One
670102 670180 96077307 670190
30617-434CS 242.21 CAD
30617-434 30617-594 30617-596 30617-592
CELLSTAR® OneWell™ and FourWell™ Plates, Greiner Bio-One
Microplates Cell Culture Plates
The new CELLSTAR® OneWell Plate™ is designed for use in applications involving the cultivation of large quantities of cells.
  • Polystyrene construction
  • Supplied with lid
  • Free of RNase, DNase, and human DNA
  • Nonpyrogenic
The external dimensions of this plate comply with ANSI standards, thus rendering the plate suitable for use with a wide range of automated cell culture and liquid handling systems
The new CELLSTAR® FourWell Plate™ allows for the cultivation and downstream processing of cells on microscopic slides formats. Designed with fourMore Product Information