ESD Safe Solvent Dispenser, R & R Lotion
Supplier: R & R Lotion
SD-6-ESD SD-6-ESD-AS
56614-114EA 81.45 CAD
56616-118 56614-114
ESD Safe Solvent Dispenser, R & R Lotion
Bottles Dropping Bottles
ESD safe solvent dispenser bottle with stainless steel anti-splash/purity pump.
  • Blue ESD container with stainless steel pump
  • Designed for use around static sensitive electronics
  • Will not hold a charge
Cleanroom blue static dissipative solvent containers have an average surface resistivity of 10⁹ to 10¹⁰. All blue solvent dispensers will dissipate a charge of +/- 5000 volts in less than 2 seconds at 40 percent relative humidity. By their chemical nature, they will not have any chemical reactivity with solvents such as TCE or alcohol. Containers help to meet ANSI/ESDA S20.20.