Support Stand, Cast Iron, Ace Glass Incorporated
Supplier: Ace Glass
13550-21 13550-23 13550-24 13550-25
89077-734EA 1293.5 CAD
89077-734 89077-736 89077-738 89077-740
Support Stand, Cast Iron, Ace Glass Incorporated
Retort Stands
Support stands feature a rugged "H" shaped, cast iron base with a 25mm O.D
stainless steel vertical rod. Stands are available in three lengths.