Zirconium(IV) acetylacetonate

Supplier: TCI America

Z0006-25G Z0006-500G Z0006-100G
TCZ0006-25GEA 72.62 CAD
TCZ0006-25G TCZ0006-500G TCZ0006-100G
Zirconium(IV) acetylacetonate
Zirconium (IV) acetylacetonate

CAS Number: 17501-44-9
MDL Number: MFCD00000036
Molecular Formula: C20H28O8Zr
Molecular Weight: 487.66
Form: Crystal
Color: White
Melting point (°C): 192
Flash Point (°C): 238


Other Product Names / Synonyms: Acetylacetone Zirconium(IV) Salt, Zirconium(IV) Acetylacetonate

Formula: C₂₀H₂₈O₈Zr
Melting Pt: 192 °C
Flash Pt: 238 °C
MDL Number: MFCD00000036
CAS Number: 17501-44-9
EINECS: 241-510-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=60252cf96431e58f99f9a2abf84d237b01bcc331&vid=60252cf96431e58f99f9a2abf84d237b01bcc331&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR