2-(Hydroxymethyl)benzyl alcohol ≥98.0%

Supplier: TCI America

Synonyms: o-Phenylenedimethanol, o-Xylene-α,α'-diol, 1,2-Benzenedimethanol

X0057-25G X0057-5G
TCX0057-25GEA 695.83 CAD
TCX0057-25G TCX0057-005G
2-(Hydroxymethyl)benzyl alcohol ≥98.0%
2-(Hydroxymethyl)benzyl alcohol

CAS Number: 612-14-6
MDL Number: MFCD00004626
Molecular Formula: C8H10O2
Molecular Weight: 138.17
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 145
Melting point (°C): 62


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: o-Xylylene Glycol, alpha,alpha'-Dihydroxy-o-xylene, Phthalyl Alcohol, o-Xylene-alpha,alpha'-diol

Formula: C₈H₁₀O₂
MW: 138.17 g/mol
Boiling Pt: 291
Melting Pt: 62 °C
MDL Number: MFCD00004626
CAS Number: 612-14-6
EINECS: 210-293-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=288dd2399522bff6b381e51f165073a97642dfdf&vid=288dd2399522bff6b381e51f165073a97642dfdf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR