Phthalan ≥97.0%

Supplier: TCI America

Synonyms: 1,3-Dihydrobenzo[c]furan

X0053-25G X0053-5G
TCX0053-025GEA 1240.3 CAD
TCX0053-025G TCX0053-005G
Phthalan ≥97.0%
Phthalan

CAS Number: 496-14-0
MDL Number: MFCD00005932
Molecular Formula: C8H8O
Molecular Weight: 120.15
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 192
Flash Point (°C): 63
Specific Gravity (20/20): 1.09


Other Product Names / Synonyms: 1,3-Dihydroisobenzofuran, Isocoumaran, Phthalan

Formula: C₈H₈O
MW: 120.15
Boiling Pt: 192 °C
Density: 1.09 (20 °C)
Flash Pt: 63 °C
MDL Number: MFCD00005932
CAS Number: 496-14-0
EINECS: 207-815-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=409636dc82969daadb2419f83ba8f2bd8db891b1&vid=409636dc82969daadb2419f83ba8f2bd8db891b1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR