Xylene cyanole FF

Supplier: TCI America
Warning

X0027-25G X0027-5G
TCX0027-025GEA 155.9 CAD
TCX0027-025G TCX0027-5G
Xylene cyanole FF
Xylene Cyanole FF

CAS Number: 2650-17-1
MDL Number: MFCD00040657
Form: Crystal
Color: Yellow Green
Lambda max.: 615 nm (H2O)


Other Product Names / Synonyms: Acid Blue 147

Formula: C₂₅H₂₇N₂NaO₆S₂
Melting Pt: 295 °C
Storage Temperature: Ambient
MDL Number: MFCD00040657
CAS Number: 2650-17-1
EINECS: 220-167-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0d0b7d21a12d701ba4acd907288f9a92abfd43f7&vid=0d0b7d21a12d701ba4acd907288f9a92abfd43f7&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR