D-(+)-Xylose 98,0
Supplier: TCI America
X0019-500G
X0019-25G
TCX0019-500GEA
205.04
CAD
TCX0019-500G
TCX0019-025G
D-(+)-Xylose 98,0
D(+)-Xylose
CAS Number: 58-86-6
MDL Number: MFCD00151475
Molecular Formula: C5H10O5
Molecular Weight: 150.13
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Color: White
Melting point (°C): 151
Specific rotation [a]20/D: 20 deg (C=10, H2O)
Formula:
C₅H₁₀O₅ MW: 150.13 g/mol Boiling Pt: 331 Melting Pt: 154 Density: 1.525 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00151475 CAS Number: 58-86-6 EINECS: 200-400-7 Merck Index: 13,10142 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...