o-Xylene ≥98.0%

Supplier: TCI America

Synonyms: 1,2-Dimethylbenzene

X0012-25ML X0012-500ML
TCX0012-25MLEA 40.91 CAD
TCX0012-500ML TCX0012-25ML
o-Xylene ≥98.0%
o-Xylene

CAS Number: 95-47-6
MDL Number: MFCD00008519
Molecular Formula: C8H10
Molecular Weight: 106.17
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 145
Melting point (°C): -23
Flash Point (°C): 33
Specific Gravity (20/20): 0.88


Other Product Names / Synonyms: 1,2-Dimethylbenzene

Formula: C₈H₁₀
MW: 106.17 g/mol
Boiling Pt: 145 °C
Melting Pt: –23 °C
Density: 0.88 (20 °C)
Flash Pt: 33 °C
MDL Number: MFCD00008519
CAS Number: 95-47-6
UN: 1307
ADR: 3,III
Merck Index: 12,10214
https://ca.vwr.com/stibo/web/std.lang.all/73/74/28997374.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR