4-Vinylphenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America

V0075-1G V0075-5G
TCV0075-1GEA 47.4 CAD
TCV0075-1G TCV0075-5G
4-Vinylphenylboronic acid (contains varying amounts of Anhydride)
4-Vinylphenylboronic acid

CAS Number: 2156-04-9
MDL Number: MFCD00239441
Molecular Formula: C8H9BO2
Molecular Weight: 147.97
Form: Crystal
Color: White


Other Product Names / Synonyms: p-Styrylboronic Acid, 4-Vinylbenzeneboronic Acid

Formula: C₈H₉BO₂
MDL Number: MFCD00239441
CAS Number: 2156-04-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fd4faee154a5a6020cce4efb02ff644b45c3a9a4&vid=fd4faee154a5a6020cce4efb02ff644b45c3a9a4&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR