4-Vinylbenzoic acid ≥97.0% stabilised

Supplier: TCI America
Warning


Stabilization: Stabilised with Stabilised

V0059-5G V0059-25G V0059-1G
TCV0059-005GEA 97.96 CAD
TCV0059-005G TCV0059-25G TCV0059-001G
4-Vinylbenzoic acid ≥97.0% stabilised
4-Vinylbenzoic acid

(stabilized with BHT)
CAS Number: 1075-49-6
MDL Number: MFCD00002569
Molecular Formula: C9H8O2
Molecular Weight: 148.16
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Color: White
Storage Temperature: 0-10°C


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: 4-Carboxystyrene

Formula: C₉H₈O₂
Boiling Pt: 282 °C (1013 hPa)
Melting Pt: 115 °C
Density: 1.3 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00002569
CAS Number: 1075-49-6
EINECS: 214-053-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=191cf027d5252bf2c878f7f7061b794860a58743&vid=10701d78a96b76d66fafdea16d7e795e92178f3d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR