(S)-(+)-2-Amino-3-methyl-1-butanol ≥97.0%

Supplier: TCI America

Synonyms: L(+)-2-Amino-3-methyl-1-butanol, (S)-2-Amino-3-methylbutan-1-ol, (S)-2-Amino-3-methyl-butan-1-ol, L-Valinol, L(+)-Valinol

V0058-25G V0058-5G
TCV0058-025GEA 470.84 CAD
TCV0058-025G TCV0058-005G
(S)-(+)-2-Amino-3-methyl-1-butanol ≥97.0%
(S)-(+)-2-Amino-3-methyl-1-butanol

CAS Number: 2026-48-4
MDL Number: MFCD00064296
Molecular Formula: C5H13NO
Molecular Weight: 103.17
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Color: White
Boiling point (°C): 88
Flash Point (°C): 91
Freezing point (°C): 33
Specific rotation [a]20/D: 17 deg (C=10, EtOH)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (S)-2-Amino-3-methyl-1-butanol

Formula: C₅H₁₃NO
Boiling Pt: 88 °C
Melting Pt: 31…32 °C
Density: 0.92 g/cm³ (20 °C)
Flash Pt: 91 °C
MDL Number: MFCD00064296
CAS Number: 2026-48-4
EINECS: 217-975-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=91decc88643caba7621b01aad31619dc96b82198&vid=91decc88643caba7621b01aad31619dc96b82198&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR