δ-Valerolactone ≥98.0%
Supplier: TCI America
V0039-25G
V0039-500G
V0039-100G
TCV0039-025GEA
95.67
CAD
TCV0039-025G
TCV0039-500G
TCV0039-100G
δ-Valerolactone ≥98.0%
δ-Valerolactone
CAS Number: 542-28-9
MDL Number: MFCD00006645
Molecular Formula: C5H8O2
Molecular Weight: 100.12
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 220
Flash Point (°C): 107
Specific Gravity (20/20): 1.11
Storage Temperature: <0°C
Product Categories: Materials Chemistry, Monomers, Bioscience, Lipids, Monomers for Synthetic Polymers (for DDS)
Other Product Names / Synonyms: Tetrahydro-2H-pyran-2-one
Formula:
C₅H₈O₂ Boiling Pt: 220 °C Melting Pt: –10 °C Density: 1.11 (20 °C) Flash Pt: 107 °C |
MDL Number:
MFCD00006645 CAS Number: 542-28-9 EINECS: 208-807-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...