Vanillic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

V0017-25G V0017-100G
TCV0017-025GEA 85.95 CAD
TCV0017-025G TCV0017-100G
Vanillic acid ≥98.0% (by GC, titration analysis)
Vanillic acid

CAS Number: 121-34-6
MDL Number: MFCD00002551
Molecular Formula: C8H8O4
Molecular Weight: 168.15
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Color: White
Melting point (°C): 210


Other Product Names / Synonyms: 4-Hydroxy-3-methoxybenzoic Acid

Formula: C₈H₈O₄
Boiling Pt: 353 °C (1013 hPa)
Melting Pt: 210 °C
Storage Temperature: Ambient
MDL Number: MFCD00002551
CAS Number: 121-34-6
EINECS: 204-466-8
Merck Index: 12,10068
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4e41d25798546e3c909f2f2b3874437933b8aa85&vid=76b795dbd90e80ff0d3da817e7acd35b04df2272&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR