L(+)-Valine ≥98.0% (by titrimetric analysis)

Supplier: TCI America

V0014-500G V0014-25G
TCV0014-500GEA 380.78 CAD
TCV0014-500G TCV0014-025G
L(+)-Valine ≥98.0% (by titrimetric analysis)
L(+)-Valine

CAS Number: 72-18-4
MDL Number: MFCD00064220
Molecular Formula: C5H11NO2
Molecular Weight: 117.15
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Specific rotation [a]20/D: 28 deg (C=8, HCl(1+1))


Product Categories: Bioscience, Proteinogenic Amino Acids
Other Product Names / Synonyms: L-2-Aminoisovaleric Acid, H-Val-OH

Formula: C₅H₁₁NO₂
Boiling Pt: 217 °C (1013 hPa)
Melting Pt: 86 °C
Density: 1.284 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064220
CAS Number: 72-18-4
EINECS: 200-773-6
Merck Index: 13,09458
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a8144a53237d5f3b1e8441e982986a56c841a9fb&vid=a8144a53237d5f3b1e8441e982986a56c841a9fb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR