DL-Valine ≥98.0% (by titrimetric analysis)
Supplier: TCI America
V0013-25G
V0013-500G
TCV0013-025GEA
45.01
CAD
TCV0013-025G
TCV0013-500G
DL-Valine ≥98.0% (by titrimetric analysis)
DL-Valine
CAS Number: 516-06-3
MDL Number: MFCD00004267
Molecular Formula: C5H11NO2
Molecular Weight: 117.15
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Melting point (°C): 295
Product Categories: Bioscience, Proteinogenic Amino Acids
Other Product Names / Synonyms: DL-2-Aminoisovaleric Acid, H-DL-Val-OH
Formula:
C₅H₁₁NO₂ Boiling Pt: 217 °C (1013 hPa) Melting Pt: 295 °C Density: 1.352 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00004267 CAS Number: 516-06-3 EINECS: 208-220-0 Merck Index: 12,10046 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...