D-(-)-Valine ≥98.0% (by titrimetric analysis)

Supplier: TCI America

V0012-25G V0012-1G
TCV0012-025GEA 231.47 CAD
TCV0012-025G TCV0012-001G
D-(-)-Valine ≥98.0% (by titrimetric analysis)
D(-)-Valine

CAS Number: 640-68-6
MDL Number: MFCD00064219
Molecular Formula: C5H11NO2
Molecular Weight: 117.15
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Specific rotation [a]20/D: -27 deg (C=8, 6mol/L HCl)


Product Categories: Bioscience, Proteinogenic Amino Acids
Other Product Names / Synonyms: D-2-Aminoisovaleric Acid, H-D-Val-OH

Formula: C₅H₁₁NO₂
Boiling Pt: 217 °C (1013 hPa)
Melting Pt: 86 °C
MDL Number: MFCD00064219
CAS Number: 640-68-6
EINECS: 211-368-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=694d68472bcd090d82791d226a4df621200746f0&vid=694d68472bcd090d82791d226a4df621200746f0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR