Valerophenone ≥98.0%

Supplier: TCI America

V0009-25ML V0009-500ML
TCV0009-025MLEA 36.34 CAD
TCV0009-025ML TCV0009-500ML
Valerophenone ≥98.0%
Valerophenone

CAS Number: 1009-14-9
MDL Number: MFCD00009480
Molecular Formula: C11H14O
Molecular Weight: 162.23
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 140
Flash Point (°C): 102
Specific Gravity (20/20): 0.98


Other Product Names / Synonyms: Butyl Phenyl Ketone, Pentanophenone

Formula: C₆H₅C(O)(CH₂)₃CH₃
Boiling Pt: 140 °C
Melting Pt: –9 °C
Density: 0.98 (20 °C)
Flash Pt: 102 °C
Storage Temperature: Ambient
MDL Number: MFCD00009480
CAS Number: 1009-14-9
EINECS: 213-767-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2584f16b192b481fbaf1d8ab1e77be6109fb67a6&vid=3bf6c22512cdad990d29bc1ffe3db3b535a55970&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR