(±)-γ-Valerolactone ≥98.0%

Supplier: TCI America

Synonyms: (±)-4-Hydroxypentanoic acid lactone, (±)-Pentan-4-olide, γ-Pentanolactone, 5-Methyldihydrofuran-2(3H)-one, gamma-Valerolactone

V0007-25G V0007-500G
TCV0007-025GEA 33.76 CAD
TCV0007-025G TCV0007-500G
(±)-γ-Valerolactone ≥98.0%
(±)-γ-Valerolactone

CAS Number: 108-29-2
MDL Number: MFCD00005400
Molecular Formula: C5H8O2
Molecular Weight: 100.12
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 207
Melting point (°C): -31
Flash Point (°C): 102
Specific Gravity (20/20): 1.06

Formula: C₅H₈O₂
MW: 100.12 g/mol
Boiling Pt: 207 °C
Melting Pt: –31 °C
Density: 1.06 (20 °C)
Flash Pt: 102 °C
MDL Number: MFCD00005400
CAS Number: 108-29-2
EINECS: 203-569-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f8f7f4f84778c5e747e496738c35ec49303ea775&vid=f8f7f4f84778c5e747e496738c35ec49303ea775&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR