Valeramide ≥98.0% (by total nitrogen basis)
Supplier: TCI America
Synonyms:
Pentanamide
V0002-25G
V0002-250G
TCV0002-025GEA
162.74
CAD
TCV0002-025G
TCV0002-250G
Valeramide ≥98.0% (by total nitrogen basis)
Valeramide
CAS Number: 626-97-1
MDL Number: MFCD00041895
Molecular Formula: C5H11NO
Molecular Weight: 101.15
Purity/Analysis Method: >98.0% (N)
Form: Crystal
Color: White
Melting point (°C): 105
Formula:
C₅H₁₀O₂ MW: 102.13 g/mol Melting Pt: 105 °C |
MDL Number:
MFCD00041895 CAS Number: 626-97-1 EINECS: 210-974-0 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...