Ethyl undecanoate ≥98.0%

Supplier: TCI America

U0049-25ML U0049-250ML
TCU0049-025MLEA 56.88 CAD
TCU0049-025ML TCU0049-250ML
Ethyl undecanoate ≥98.0%
Ethyl undecanoate

CAS Number: 627-90-7
MDL Number: MFCD00008958
Molecular Formula: C13H26O2
Molecular Weight: 214.35
Purity/Analysis Method: >98.0% (GC)
Boiling point (°C): 140
Flash Point (°C): 113
Specific Gravity (20/20): 0.87


Product Categories: Bioscience, Lipids
Other Product Names / Synonyms: Undecanoic Acid Ethyl Ester

Formula: C₁₃H₂₆O₂
Boiling Pt: 140 °C
Melting Pt: –14.7 °C
Density: 0.87 (20 °C)
Flash Pt: 113 °C
MDL Number: MFCD00008958
CAS Number: 627-90-7
EINECS: 211-018-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=922d4630142979b79c477abfbc8ff6bcf06c234b&vid=922d4630142979b79c477abfbc8ff6bcf06c234b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR