10-Undecen-1-ol ≥98.0%
Supplier: TCI America
U0047-25ML
U0047-500ML
TCU0047-025MLEA
65.89
CAD
TCU0047-025ML
TCU0047-500ML
10-Undecen-1-ol ≥98.0%
10-Undecen-1-ol
CAS Number: 112-43-6
MDL Number: MFCD00004750
Molecular Formula: C11H22O
Molecular Weight: 170.30
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 138
Flash Point (°C): 110
Specific Gravity (20/20): 0.85
Formula:
C₁₁H₂₂O Boiling Pt: 138 °C Melting Pt: –1 °C Density: 0.85 (20 °C) Flash Pt: 110 °C |
MDL Number:
MFCD00004750 CAS Number: 112-43-6 EINECS: 203-971-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...