Uracil ≥98.0%

Supplier: TCI America

Synonyms: Pyrimidine-2,4(1H,3H)-dione

U0013-500G U0013-25G U0013-100G
TCU0013-500GEA 265.44 CAD
TCU0013-500G TCU0013-025G TCU0013-100G
Uracil ≥98.0%
Uracil

CAS Number: 66-22-8
MDL Number: MFCD00006016
Molecular Formula: C4H4N2O2
Molecular Weight: 112.09
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Color: White
Melting point (°C): 335


Product Categories: Bioscience, Nucleobases and Their Analogs
Other Product Names / Synonyms: 2,4(1H,3H)-Pyrimidinedione

Formula: C₄H₄N₂O₂
MW: 112.09 g/mol
Boiling Pt: 379
Melting Pt: 194
Density: 0.99744 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006016
CAS Number: 66-22-8
EINECS: 200-621-9
Merck Index: 12,09985
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0205dd888f415bc44387cd510365e2633d90ae07&vid=0205dd888f415bc44387cd510365e2633d90ae07&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR