(±)-γ-Undecalactone ≥98.0%

Supplier: TCI America

U0003-25ML U0003-500ML
TCU0003-025MLEA 26.86 CAD
TCU0003-025ML TCU0003-500ML
(±)-γ-Undecalactone ≥98.0%
(±)-γ-Undecalactone

CAS Number: 104-67-6
MDL Number: MFCD00005405
Molecular Formula: C11H20O2
Molecular Weight: 184.28
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 110
Flash Point (°C): 156
Specific Gravity (20/20): 0.95

Formula: C₁₁H₂₀O₂
Boiling Pt: 110 °C
Density: 0.95 (20 °C)
Flash Pt: 156 °C
MDL Number: MFCD00005405
CAS Number: 104-67-6
EINECS: 203-225-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ca9d2565175ac1412dfc086b21863e30bcae68f5&vid=ca9d2565175ac1412dfc086b21863e30bcae68f5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR