Tris(hydroxymethyl)aminomethane (TRIS, Trometamol) ≥99.0%
Supplier: TCI America
|
Warning
|
Synonyms:
2-Amino-2-(hydroxymethyl)propane-1,3-diol(tris), Tromethamine, Tris(hydroxymethyl)methylamine, 2-Amino-2-(hydroxymethyl)propane-1,3-diol, 2-Amino-2-(hydroxymethyl)-1,3-propanediol, Trizma(R) base, TRIS base, THAM
CAS Number: 77-86-1
MDL Number: MFCD00004679
Molecular Formula: C4H11NO3
Molecular Weight: 121.14
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Color: White
Boiling point (°C): 220
Melting point (°C): 170
Product Categories: Bioscience, Buffer-related Reagents, Analytical Science, Reagents for Gel & Buffer Preparation, etc. (Reagents for Electrophoresis)
Other Product Names / Synonyms: 2-Amino-2-(hydroxymethyl)-1,3-propanediol, Trimethylolaminomethane, Tris-Base
Formula:
NH₂C(CH₂OH)₃ MW: 121.14 g/mol Boiling Pt: 357 Melting Pt: 170 °C Density: 1.328 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00004679 CAS Number: 77-86-1 EINECS: 201-064-4 Merck Index: 13,09842 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...