2,4-Thiazolidinedione ≥98.0% (by GC, titration analysis)
Supplier: TCI America
T1990-25G
T1990-500G
T1990-100G
TCT1990-25GEA
30.02
CAD
TCT1990-25G
TCT1990-500G
76220-514
2,4-Thiazolidinedione ≥98.0% (by GC, titration analysis)
2,4-Thiazolidinedione
CAS Number: 2295-31-0
MDL Number: MFCD00005478
Molecular Formula: C3H3NO2S
Molecular Weight: 117.12
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Color: White
Melting point (°C): 128
Formula:
C₃H₃NO₂S Melting Pt: 128 °C |
MDL Number:
MFCD00005478 CAS Number: 2295-31-0 EINECS: 218-941-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...