3-(Trifluoromethoxy)benzaldehyde ≥96.0%

Supplier: TCI America
Warning

T1824-25G T1824-5G
TCT1824-25GEA 540.36 CAD
TCT1824-25G TCT1824-005G
3-(Trifluoromethoxy)benzaldehyde ≥96.0%
3-(Trifluoromethoxy)benzaldehyde

CAS Number: 52771-21-8
MDL Number: MFCD00042406
Molecular Formula: C8H5F3O2
Molecular Weight: 190.12
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Color: Colorless
Flash Point (°C): 73
Specific Gravity (20/20): 1.33

Formula: C₈H₅F₃O₂
Boiling Pt: 83…86 °C (24 mmHg)
Melting Pt: 450 °C
Density: 1.33 (20 °C)
Flash Pt: 73 °C
MDL Number: MFCD00042406
CAS Number: 52771-21-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4afa2814eab6094393102e0839a5cf1f8054b3f7&vid=4afa2814eab6094393102e0839a5cf1f8054b3f7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR