(±)-Tetrahydrofurfuryl bromide ≥95.0%
Supplier: TCI America
T1808-500G
T1808-25G
T1808-100G
TCT1808-500GEA
499.28
CAD
TCT1808-500G
TCT1808-025G
TCT1808-100G
(±)-Tetrahydrofurfuryl bromide ≥95.0%
(±)-Tetrahydrofurfuryl bromide
CAS Number: 1192-30-9
MDL Number: MFCD00016894
Molecular Formula: C5H9BrO
Molecular Weight: 165.03
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 70
Flash Point (°C): 73
Specific Gravity (20/20): 1.47
Storage Temperature: 0-10°C
Other Product Names / Synonyms: 2-(Bromomethyl)tetrahydrofuran
Formula:
C₅H₉BrO Boiling Pt: 70 °C Density: 1.47 (20 °C) Flash Pt: 73 °C |
MDL Number:
MFCD00016894 CAS Number: 1192-30-9 EINECS: 214-750-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...