4-(Trifluoromethoxy)phenylacetonitrile ≥98.0%

Supplier: TCI America

Synonyms: 1-(Cyanmethyl)-4-(trifluormethoxy)benzol, 4-(TrifluoroMethoxy)phenylacetonitrile, 4-(Trifluoromethoxy)benzyl cyanide

T1804-5G T1804-1G
TCT1804-005GEA 133.83 CAD
TCT1804-005G TCT1804-001G
4-(Trifluoromethoxy)phenylacetonitrile ≥98.0%
4-(Trifluoromethoxy)phenylacetonitrile

CAS Number: 49561-96-8
MDL Number: MFCD00061240
Molecular Formula: C9H6F3NO
Molecular Weight: 201.15
Purity/Analysis Method: >98.0% (GC)
Boiling point (°C): 96
Flash Point (°C): 113
Freezing point (°C): 27

Formula: C₉H₆F₃NO
MW: 201.15 g/mol
Boiling Pt: 96 °C
Melting Pt: 28…30 °C
Density: 1.28 g/cm³ (20 °C)
Flash Pt: 113 °C
MDL Number: MFCD00061240
CAS Number: 49561-96-8
UN: 3439
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=92940696701904b9ec2955c7a2521d42ce7d83cf&vid=92940696701904b9ec2955c7a2521d42ce7d83cf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR