2-(Trifluoromethyl)phenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America

Synonyms: (2-Trifluoromethyl)phenylboronic acid, (2-(Trifluoromethyl)phenyl)metaboric acid, Dihydroxy(2-(trifluoromethyl)phenyl)borane, 2-(Trifluoromethyl)benzeneboronic acid

T1800-1G T1800-5G T1800-25G
TCT1800-001GEA 48.98 CAD
TCT1800-001G TCT1800-005G TCT1800-25G
2-(Trifluoromethyl)phenylboronic acid (contains varying amounts of Anhydride)
2-(Trifluoromethyl)phenylboronic acid

CAS Number: 1423-27-4
MDL Number: MFCD00236059
Molecular Formula: C7H6BF3O2
Molecular Weight: 189.93
Form: Crystal
Color: White


Other Product Names / Synonyms: alpha,alpha,alpha-Trifluoro-o-tolueneboronic Acid, alpha,alpha,alpha-Trifluoro-o-tolylboronic Acid

Formula: C₇H₆BF₃O₂
MW: 189.93 g/mol
MDL Number: MFCD00236059
CAS Number: 1423-27-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1283ee80b4ff46ca5363608d4079ce9dbe17b620&vid=1283ee80b4ff46ca5363608d4079ce9dbe17b620&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR