3-(Trifluoromethyl)phenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America

Synonyms: (3-(Trifluoromethyl)phenyl)boronic acid, (3-(Trifluoromethyl)phenyl)metaboric acid, 3-(Trifluoromethyl)benzeneboronic acid, Dihydroxy(3-(trifluoromethyl)phenyl)borane

T1793-1G T1793-5G
TCT1793-001GEA 33.18 CAD
TCT1793-001G TCT1793-5G
3-(Trifluoromethyl)phenylboronic acid (contains varying amounts of Anhydride)
3-(Trifluoromethyl)phenylboronic acid

CAS Number: 1423-26-3
MDL Number: MFCD00151854
Molecular Formula: C7H6BF3O2
Molecular Weight: 189.93
Form: Crystal
Color: White


Other Product Names / Synonyms: alpha,alpha,alpha-Trifluoro-m-tolueneboronic Acid, alpha,alpha,alpha-Trifluoro-m-tolylboronic Acid

Formula: C₇H₆BF₃O₂
MW: 189.93 g/mol
MDL Number: MFCD00151854
CAS Number: 1423-26-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f7b98a61752eb7c8c0582c53e6e51d5324895bbe&vid=f7b98a61752eb7c8c0582c53e6e51d5324895bbe&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR