4-(Trifluoromethyl)phenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America

Synonyms: 4-(Trifluoromethyl)benzeneboronic acid, (4-(Trifluoromethyl)phenyl)metaboric acid, Dihydroxy(4-(trifluoromethyl)phenyl)borane, (4-(Trifluoromethyl)phenyl)boronic acid

T1788-5G T1788-1G T1788-25G
TCT1788-5GEA 227.52 CAD
TCT1788-5G TCT1788-001G TCT1788-25G
4-(Trifluoromethyl)phenylboronic acid (contains varying amounts of Anhydride)
4-(Trifluoromethyl)phenylboronic acid

CAS Number: 128796-39-4
MDL Number: MFCD00151855
Molecular Formula: C7H6BF3O2
Molecular Weight: 189.93
Form: Crystal
Color: White
Melting point (°C): 250


Other Product Names / Synonyms: alpha,alpha,alpha-Trifluoro-p-tolueneboronic Acid, alpha,alpha,alpha-Trifluoro-p-tolylboronic Acid

Formula: C₇H₆BF₃O₂
MW: 189.93 g/mol
Melting Pt: 250 °C
MDL Number: MFCD00151855
CAS Number: 128796-39-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=95f5268a0723beb4bac4d0c0d74f8f7e35610be7&vid=95f5268a0723beb4bac4d0c0d74f8f7e35610be7&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR