4'-(Trifluoromethyl)acetophenone ≥98.0%

Supplier: TCI America
Warning

T1784-25G T1784-1G T1784-5G
TCT1784-25GEA 247.9 CAD
TCT1784-25G TCT1784-001G TCT1784-005G
4'-(Trifluoromethyl)acetophenone ≥98.0%
4'-(Trifluoromethyl)acetophenone

CAS Number: 709-63-7
MDL Number: MFCD00000401
Molecular Formula: C9H7F3O
Molecular Weight: 188.15
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 80
Flash Point (°C): 84
Freezing point (°C): 29


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 4-Acetylbenzotrifluoride

Formula: C₉H₇F₃O
Boiling Pt: 80 °C
Melting Pt: 26…29 °C
Density: 0.924 g/cm³ (20 °C)
Flash Pt: 84 °C
Storage Temperature: Ambient
MDL Number: MFCD00000401
CAS Number: 709-63-7
EINECS: 211-913-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5c41c1802eb7d179d15c6cef2d61ad7f1a0b6c00&vid=5c41c1802eb7d179d15c6cef2d61ad7f1a0b6c00&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR