4-(Trifluoromethoxy)phenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America

Synonyms: Dihydroxy(4-(trifluoromethoxy)phenyl)borane, (4-(Trifluoromethoxy)phenyl)metaboric acid, 4-(Trifluoromethoxy)benzeneboronic acid, 4-Trifluoromethoxyphenylboronic acid

T1773-5G T1773-25G
TCT1773-005GEA 83.74 CAD
TCT1773-005G TCT1773-25G
4-(Trifluoromethoxy)phenylboronic acid (contains varying amounts of Anhydride)
4-(Trifluoromethoxy)phenylboronic acid

CAS Number: 139301-27-2
MDL Number: MFCD01074648
Molecular Formula: C7H6BF3O3
Molecular Weight: 205.93
Form: Crystal
Color: White
Melting point (°C): 127


Other Product Names / Synonyms: 4-(Trifluoromethoxy)benzeneboronic Acid

Formula: C₇H₆BF₃O₃
MW: 205.93 g/mol
Melting Pt: 127 °C
MDL Number: MFCD01074648
CAS Number: 139301-27-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=58ea877be17d420db04973cc7b419a2d24f9aeb4&vid=58ea877be17d420db04973cc7b419a2d24f9aeb4&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR