2-Thiopheneboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America

Synonyms: Thiophen-2-ylboronic acid, Thiophene-2-boronic acid, 2-Thienylboronic acid

T1772-25G T1772-5G T1772-1G
TCT1772-25GEA 383.94 CAD
TCT1772-25G TCT1772-5G TCT1772-001G
2-Thiopheneboronic acid (contains varying amounts of Anhydride)
2-Thiopheneboronic acid

CAS Number: 6165-68-0
MDL Number: MFCD00151850
Molecular Formula: C4H5BO2S
Molecular Weight: 127.95
Form: Crystal
Color: White
Storage Temperature: 0-10°C


Other Product Names / Synonyms: 2-Thienylboronic Acid

Formula: C₄H₅BO₂S
MW: 127.96 g/mol
Melting Pt: 146…147 °C
MDL Number: MFCD00151850
CAS Number: 6165-68-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6271500781c52d87300bac0ae6ac95a9270ff207&vid=6271500781c52d87300bac0ae6ac95a9270ff207&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR