Trimethylsulfonium bromide ≥98.0%

Supplier: TCI America

Synonyms: Trimethylsulfoniumbromide, Trimethylsulphoniumbromide

T1771-25G T1771-100G
TCT1771-025GEA 79 CAD
TCT1771-025G TCT1771-100G
Trimethylsulfonium bromide ≥98.0%
Trimethylsulphonium bromide

CAS Number: 3084-53-5
MDL Number: MFCD00051367
Molecular Formula: C3H9BrS
Molecular Weight: 157.07
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Flash Point (°C): 110

Formula: C₃H₉BrS
MW: 157.07 g/mol
Melting Pt: 203
Density: 1.67 g/cm³ (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00051367
CAS Number: 3084-53-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eec675f6833ae147654b96778f835e3420c784e3&vid=eec675f6833ae147654b96778f835e3420c784e3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR