2,4,5-Trihydroxybenzaldehyde ≥98.0%

Supplier: TCI America
Warning

T1766-1G T1766-5G
TCT1766-001GEA 188.49 CAD
TCT1766-001G TCT1766-5G
2,4,5-Trihydroxybenzaldehyde ≥98.0%
2,4,5-Trihydroxybenzaldehyde

CAS Number: 35094-87-2
MDL Number: MFCD00016592
Molecular Formula: C7H6O4
Molecular Weight: 154.12
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: Yellow

Formula: C₇H₆O₄
Melting Pt: 228 °C
MDL Number: MFCD00016592
CAS Number: 35094-87-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f09ac09274118993f94beffb1834e2fc26d3bc34&vid=94637f28fb20439f140df28b7d2efd45164d8c84&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR