Tetrafluoroisophthalonitrile ≥98.0%

Supplier: TCI America

Synonyms: Tetrafluoro-1,3-benzenedicarbonitrile, Tetrafluorobenzene-1,3-dicarbonitrile, Tetrafluoro-1,3-dicyanobenzene

T1728-25G T1728-5G
TCT1728-025GEA 381.89 CAD
TCT1728-025G TCT1728-005G
Tetrafluoroisophthalonitrile ≥98.0%
Tetrafluoroisophthalonitrile

CAS Number: 2377-81-3
MDL Number: MFCD00013290
Molecular Formula: C8F4N2
Molecular Weight: 200.10
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 76


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: Perfluoroisophthalonitrile

Formula: C₈F₄N₂
MW: 200.1 g/mol
Boiling Pt: 283 °C (1013 hPa)
Melting Pt: 76 °C
MDL Number: MFCD00013290
CAS Number: 2377-81-3
UN: 3439
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=205bb738d2419cb4ecaa061d8760540488cde928&vid=205bb738d2419cb4ecaa061d8760540488cde928&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR