2,3,5-Collidine ≥98.0%

Supplier: TCI America
Warning

Synonyms: 2,3,5-Trimethylpyridine

T1722-25G T1722-5G
TCT1722-25GEA 94.8 CAD
TCT1722-25G TCT1722-005G
2,3,5-Collidine ≥98.0%
2,3,5-Collidine

CAS Number: 695-98-7
MDL Number: MFCD00126840
Molecular Formula: C8H11N
Molecular Weight: 121.18
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 65
Flash Point (°C): 74
Specific Gravity (20/20): 0.94

Formula: C₈H₁₁N
MW: 121.18 g/mol
Boiling Pt: 65 °C
Density: 0.94 (20 °C)
Flash Pt: 74 °C
MDL Number: MFCD00126840
CAS Number: 695-98-7
EINECS: 211-786-1
UN: 2810
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fb45952f6181abd7df14944e65e2edc9ace13774&vid=fb45952f6181abd7df14944e65e2edc9ace13774&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR