2,4,6-Tri-tert-butylaniline ≥98.0%

Supplier: TCI America
Warning

Synonyms: 2,4,6-Tris(tert-butyl)aniline

T1694-1G T1694-5G
TCT1694-001GEA 71.1 CAD
TCT1694-001G TCT1694-005G
2,4,6-Tri-tert-butylaniline ≥98.0%
2,4,6-Tri-tert-butylaniline

CAS Number: 961-38-6
MDL Number: MFCD00011645
Molecular Formula: C18H31N
Molecular Weight: 261.45
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Color: White
Melting point (°C): 147

Formula: C₁₈H₃₁N
Boiling Pt: 302 °C (1013 hPa)
Melting Pt: 147 °C
Density: 0.991 g/cm³ (19.85 °C)
MDL Number: MFCD00011645
CAS Number: 961-38-6
EINECS: 213-507-9
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eb39554c276399de048fa4db57b63e87dc5d1557&vid=1e6b99bc46e6e2669fc9397a264b1025bed2474e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR