1,3,5-Trifluoro-2-nitrobenzene ≥97.0%

Supplier: TCI America

T1681-5G T1681-25G
TCT1681-005GEA 115.34 CAD
TCT1681-005G TCT1681-25G
1,3,5-Trifluoro-2-nitrobenzene ≥97.0%
1,3,5-Trifluoro-2-nitrobenzene

CAS Number: 315-14-0
MDL Number: MFCD00014687
Molecular Formula: C6H2F3NO2
Molecular Weight: 177.08
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Color: Slightly Pale Yellow
Boiling point (°C): 172
Flash Point (°C): 78
Specific Gravity (20/20): 1.53


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 1,3,5-Trifluoro-2-nitrobenzene

Formula: C₆H₂F₃NO₂
Boiling Pt: 172 °C
Melting Pt: 3.5 °C
Density: 1.53 (20 °C)
Flash Pt: 78 °C
MDL Number: MFCD00014687
CAS Number: 315-14-0
EINECS: 206-248-8
UN: 2810
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=94bbbe2995f6c2edb2b2853367427207f91c75a0&vid=079485079815429df669fbba30aafff283331045&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR