1-Bromo-2,3,4-trifluorobenzene ≥98.0%
Supplier: TCI America
|
Warning
|
T1678-5G
T1678-1G
TCT1678-005GEA
123.24
CAD
TCT1678-005G
TCT1678-001G
1-Bromo-2,3,4-trifluorobenzene ≥98.0%
1-Bromo-2,3,4-trifluorobenzene
CAS Number: 176317-02-5
MDL Number: MFCD00013248
Molecular Formula: C6H2BrF3
Molecular Weight: 210.98
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 60
Flash Point (°C): 62
Specific Gravity (20/20): 1.81
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 2,3,4-Trifluorobromobenzene
Formula:
C₆H₂BrF₃ Boiling Pt: 60 °C Density: 1.81 (20 °C) Flash Pt: 62 °C |
MDL Number:
MFCD00013248 CAS Number: 176317-02-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...