1,2,3,4-Tetrahydroisoquinoline ≥95.0%
Supplier: TCI America
T1676-500G
T1676-25G
T1676-100G
TCT1676-500GEA
575.12
CAD
TCT1676-500G
TCT1676-025G
TCT1676-100G
1,2,3,4-Tetrahydroisoquinoline ≥95.0%
1,2,3,4-Tetrahydroisoquinoline
CAS Number: 91-21-4
MDL Number: MFCD00006896
Molecular Formula: C9H11N
Molecular Weight: 133.19
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 233
Melting point (°C): -30
Flash Point (°C): 110
Specific Gravity (20/20): 1.05
Formula:
C₉H₁₁N MW: 133.19 g/mol Boiling Pt: 233 °C Melting Pt: –30 °C Density: 1.05 (20 °C) Flash Pt: 110 °C |
MDL Number:
MFCD00006896 CAS Number: 91-21-4 EINECS: 202-050-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...