(α,α,α-Trifluoro-2-tolyl)acetic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

T1661-5G T1661-25G T1661-1G
TCT1661-005GEA 71.1 CAD
TCT1661-005G TCT1661-25G TCT1661-1G
(α,α,α-Trifluoro-2-tolyl)acetic acid ≥98.0% (by GC, titration analysis)
(α,α,α-Trifluoro-2-tolyl)acetic acid

CAS Number: 3038-48-0
MDL Number: MFCD00004327
Molecular Formula: C9H7F3O2
Molecular Weight: 204.15
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Color: White
Melting point (°C): 102


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: (alpha,alpha,alpha-Trifluoro-o-tolyl)acetic Acid

Formula: F₃CC₆H₄CH₂COOH
Boiling Pt: 272 °C (1013 hPa)
Melting Pt: 102 °C
MDL Number: MFCD00004327
CAS Number: 3038-48-0
EINECS: 221-240-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9457d6b57b71ca7dbf9ae0904dc861c114b5c6ab&vid=9457d6b57b71ca7dbf9ae0904dc861c114b5c6ab&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR