(-)-Dimethyl-D-tartrate ≥98.0%

Supplier: TCI America

Synonyms: D(-)-Tartaric acid dimethyl ester, (2S,3S)-Dimethyl 2,3-dihydroxysuccinate, (S,S)-(-)-Tartaric acid dimethyl ester, (S,S)-(-)-Dimethyl tartrate, (-)-Dimethyl D-tartrate

T1659-5G T1659-25G
TCT1659-005GEA 112.18 CAD
TCT1659-005G TCT1659-025G
(-)-Dimethyl-D-tartrate ≥98.0%
(-)-Dimethyl D-tartrate

CAS Number: 13171-64-7
MDL Number: MFCD00008445
Molecular Formula: C6H10O6
Molecular Weight: 178.14
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 118
Melting point (°C): 60
Flash Point (°C): 110
Specific rotation [a]20/D: -21 deg (C=1, H2O)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: D-(-)-Tartaric Acid Dimethyl Ester

Formula: C₆H₁₀O₆
MW: 178.14 g/mol
Boiling Pt: 118 °C
Melting Pt: 60 °C
Flash Pt: 110 °C
MDL Number: MFCD00008445
CAS Number: 13171-64-7
EINECS: 236-118-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9b919744190e338eb6d965c99843cd5fb03615aa&vid=9b919744190e338eb6d965c99843cd5fb03615aa&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR