3,4,5-Trifluorobenzoic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

T1640-1G T1640-5G
TCT1640-1GEA 44.24 CAD
TCT1640-1G TCT1640-005G
3,4,5-Trifluorobenzoic acid ≥98.0% (by titrimetric analysis)
3,4,5-Trifluorobenzoic acid

CAS Number: 121602-93-5
MDL Number: MFCD00074962
Molecular Formula: C7H3F3O2
Molecular Weight: 176.09
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Melting point (°C): 98

Formula: C₇H₃F₃O₂
Boiling Pt: 237 °C (1013 hPa)
Melting Pt: 98 °C
MDL Number: MFCD00074962
CAS Number: 121602-93-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1fb9822f59851c998126df6d00131ba185407823&vid=7ebee7c74be86d29f99ba00875e063f405a2f8c4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR