Tetrafluorosuccinic acid ≥97.0% (by titrimetric analysis)

Supplier: TCI America
Warning

Synonyms: 2,2,3,3-Tetrafluorosuccinic acid, Tetrafluoro-1,4-butanedioic acid

T1621-25G T1621-5G
TCT1621-025GEA 398.16 CAD
TCT1621-025G TCT1621-005G
Tetrafluorosuccinic acid ≥97.0% (by titrimetric analysis)
Tetrafluorosuccinic acid

CAS Number: 377-38-8
MDL Number: MFCD00042430
Molecular Formula: C4H2F4O4
Molecular Weight: 190.05
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Color: White
Boiling point (°C): 120
Melting point (°C): 116


Other Product Names / Synonyms: Perfluoro-1,4-butanedioic Acid, Perfluorosuccinic Acid, Tetrafluoro-1,4-butanedioic Acid

Formula: C₄H₂F₄O₄
MW: 190.05 g/mol
Boiling Pt: 120 °C
Melting Pt: 116 °C
Density: 1.4 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00042430
CAS Number: 377-38-8
EINECS: 206-820-7
UN: 3261
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=32facae804993b37f2c7121b56f481ac1806e9b6&vid=32facae804993b37f2c7121b56f481ac1806e9b6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR