3-(Trifluoromethoxy)phenol ≥95.0%
Supplier: TCI America
Synonyms:
α,α,α-Trifluoro-3-hydroxyanisole
T1615-5G
T1615-25G
TCT1615-005GEA
102.7
CAD
TCT1615-005G
TCT1615-25G
3-(Trifluoromethoxy)phenol ≥95.0%
3-(Trifluoromethoxy)phenol
CAS Number: 827-99-6
MDL Number: MFCD00040987
Molecular Formula: C7H5F3O2
Molecular Weight: 178.11
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 80
Flash Point (°C): 84
Specific Gravity (20/20): 1.37
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 3-Hydroxyphenyl Trifluoromethyl Ether
Formula:
C₇H₅F₃O₂ MW: 178.11 g/mol Boiling Pt: 80 °C Density: 1.37 (20 °C) Flash Pt: 84 °C |
MDL Number:
MFCD00040987 CAS Number: 827-99-6 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...