1,4,8,11-Tetraazacyclotetradecane ≥98.0% (by titrimetric analysis)

Supplier: TCI America

T1597-1G T1597-5G
TCT1597-001GEA 145.36 CAD
TCT1597-001G TCT1597-005G
1,4,8,11-Tetraazacyclotetradecane ≥98.0% (by titrimetric analysis)
1,4,8,11-Tetraazacyclotetradecane

CAS Number: 295-37-4
MDL Number: MFCD00005105
Molecular Formula: C10H24N4
Molecular Weight: 200.33
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Melting point (°C): 187


Product Categories: Materials Chemistry, Precise Polymerization Reagents, Macrocycles for Host-Guest Chemistry, Bioscience, Polyamines
Other Product Names / Synonyms: Cyclam

Formula: C₁₀H₂₄N₄
Melting Pt: 187 °C
MDL Number: MFCD00005105
CAS Number: 295-37-4
EINECS: 206-039-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8bdc17754b7e98a4516744dfaaf07c95816f33d3&vid=8bdc17754b7e98a4516744dfaaf07c95816f33d3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR